6-Bromo-4-chloroquinoline 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR43545-1G
OR43545-5G
OR43545-10G
76583-804EA
35
USD
76583-804
76583-806
76583-802
6-Bromo-4-chloroquinoline 98%
6-Bromo-4-chloroquinoline
Formula:
C₉H₅BrClN MW: 242.50 g/mol Melting Pt: 123…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00511001 CAS Number: 65340-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...