Methyl-4-iodo-3-methylbenzoate 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 4-iodo-3-methylbenzoate

OR4700-100G OR4700-5G OR4700-25G
76585-322EA 109 USD
76585-322 76585-324 76585-320
Methyl-4-iodo-3-methylbenzoate 97%
Methyl 4-iodo-3-methylbenzoate
Formula: C₉H₉IO₂
MW: 276.07 g/mol
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD02683863
CAS Number: 5471-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR