Pyridazine-4-carboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Pyridazinecarboxylic acid

OR7883-1G OR7883-250MG OR7883-100MG
76588-366EA 46 USD
76588-366 76588-364 76588-362
Pyridazine-4-carboxylic acid
Pyridazine-4-carboxylic acid
Formula: C₅H₄N₂O₂
MW: 124.10 g/mol
Melting Pt: 244 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011576
CAS Number: 50681-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR