5-Amino-3-bromo-1-methyl-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Bromo-1-methyl-1H-indazol-5-amine

OR43550-1G OR43550-250MG
76583-810EA 18 USD
76583-810 76583-812
5-Amino-3-bromo-1-methyl-1H-indazole
5-Amino-3-bromo-1-methyl-1H-indazole
Formula: C₈H₈BrN₃
MW: 226.0700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11100026
CAS Number: 1092351-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR