7-Bromo-1-methyl-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 7-Bromo-1-methylindazole

OR43560-250MG OR43560-1G
76583-826EA 43 USD
76583-826 76583-828
7-Bromo-1-methyl-1H-indazole
7-Bromo-1-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878570
CAS Number: 1000576-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR