6-Bromo-2-methyl-2H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

OR43559-250MG OR43559-1G
76583-822EA 13.39 USD
76583-822 76583-824
6-Bromo-2-methyl-2H-indazole
6-Bromo-2-methyl-2H-indazole
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09870048
CAS Number: 590417-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR