(5-Methyl-1H-pyrazol-1-yl)acetic acid

Supplier: APOLLO SCIENTIFIC

Warning

OR451406-5G OR451406-1G
76584-388EA 112 USD
76584-388 76584-386
(5-Methyl-1H-pyrazol-1-yl)acetic acid
(5-Methyl-1H-pyrazol-1-yl)acetic acid
Formula: C₆H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03419599
CAS Number: 180741-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR