4-Bromo-2-(methylsulfonyl)benzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR7823-1G
76588-300EA 48 USD
76588-300
4-Bromo-2-(methylsulfonyl)benzoic acid
4-Bromo-2-(methylsulphonyl)benzoic acid
Formula: C₈H₇BrO₄S
Melting Pt: 220…224 °C
Storage Temperature: Ambient
MDL Number: MFCD04037926
CAS Number: 142994-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR