1-Bromo-2-chloro-5-nitrobenzene 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Bromo-4-chloronitrobenzene

OR4221-500G OR4221-100G OR4221-25G
76583-460EA 55 USD
76583-460 76583-462 76583-464
1-Bromo-2-chloro-5-nitrobenzene 98%
1-Bromo-2-chloro-5-nitrobenzene
Formula: C₆H₃BrClNO₂
MW: 236.4500 g/mol
Melting Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00100437
CAS Number: 16588-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR