1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR41018-5G OR41018-25G OR41018-100G
76583-196EA 35 USD
76583-196 76583-198 76583-200
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester 97%
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₄H₂₃BN₂O₃
MW: 278.1550 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16556148
CAS Number: 1003846-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR