2-Bromo-4-chloro-6-nitroaniline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-4-chloro-6-nitrophenylamine

OR7021-25G OR7021-100G OR7021-5G
76587-554EA 13.39 USD
76587-554 76587-556 76587-558
2-Bromo-4-chloro-6-nitroaniline
2-Bromo-4-chloro-6-nitroaniline
Formula: C₆H₄BrClN₂O₂
MW: 251.4700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08275709
CAS Number: 827-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR