Benzyloxyacetaldehyde diethyl acetal 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR451328-10G OR451328-50G
76584-338EA 42 USD
76584-338 76584-340
Benzyloxyacetaldehyde diethyl acetal 95%
Benzyloxyacetaldehyde diethyl acetal
Formula: C₁₃H₂₀O₃
MW: 224.30 g/mol
Boiling Pt: 99…100 °C
Density: 0.987 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00051786
CAS Number: 42783-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR