4-Benzyloxybenzophenone 98+%

Supplier: APOLLO SCIENTIFIC
Warning

OR451318-25G OR451318-5G OR451318-100G
76584-318EA 69 USD
76584-318 76584-316 76584-320
4-Benzyloxybenzophenone 98+%
4-Benzyloxybenzophenone
Formula: C₁₄H₉F₃O
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00017508
CAS Number: 54589-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR