5-Pyrimidineboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Pyrimidinylboronic acidPyrimidin-5-ylboronic acidPyrimidine-5-boronic acid

OR7262-10G OR7262-1G OR7262-5G
76587-840EA 31 USD
76587-840 76587-842 76587-838
5-Pyrimidineboronic acid
5-Pyrimidineboronic acid
Formula: C₄H₅BN₂O₂
MW: 123.91 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03002366
CAS Number: 109299-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR