4-(Heptyloxy)phenylboronic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-n-Heptyloxybenzeneboronic acid4-Heptyloxyphenylboronic acid4-Heptyloxybenzeneboronic acid

OR7221-25G OR7221-1G OR7221-5G
76587-784EA 13 USD
76587-784 76587-786 76587-788
4-(Heptyloxy)phenylboronic acid
4-(Heptyloxy)phenylboronic acid
Formula: C₁₃H₂₁BO₃
MW: 236.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039169
CAS Number: 136370-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR