Tetrahydropyran-3-methanol

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-(Hydroxymethyl)tetrahydro-2H-pyran(Tetrahydropyran-3-yl)methanol

OR70047-1G OR70047-5G
76587-410EA 359 USD
76587-410 76587-408
Tetrahydropyran-3-methanol
Tetrahydropyran-3-methanol
Formula: C₆H₁₂O₂
MW: 116.1583 g/mol
Boiling Pt: 81 °C/4mmHg
Density: 1.0553 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD08235011
CAS Number: 14774-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR