2-Chloro-5-methylphenylboronic acid 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2-Chloro-5-methylphenyl)boronic acid2-Chloro-5-methylbenzeneboronic acid

OR4249-1G OR4249-5G
76583-628EA 13.39 USD
76583-628 76583-630
2-Chloro-5-methylphenylboronic acid 96%
2-Chloro-5-methylphenylboronic acid
Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Melting Pt: 203…205 °C
Storage Temperature: Ambient
MDL Number: MFCD06659822
CAS Number: 193353-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR