Ethyl-2-methylnicotinate 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Ethyl 2-methylnicotinate

OR42000-5G OR42000-1G
76583-340EA 19 USD
76583-340 76583-338
Ethyl-2-methylnicotinate 98%
Ethyl 2-methylnicotinate
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 126…127 °C/24mmHg
Density: 1.072 g/cm³ (25 °C)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006336
CAS Number: 1721-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR