4-(Isobutyro)thioanisole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: P-(methylthio)isobutyrophenone2-Methyl-1-[4-(methylsulphanyl)phenyl]propan-1-one

OR4659-100G OR4659-25G
76585-134EA 24 USD
76585-134 76585-136
4-(Isobutyro)thioanisole
4-(Isobutyro)thioanisole
Formula: C₁₁H₁₄OS
MW: 194.2933 g/mol
Boiling Pt: 122…124 °C/2mmHg
Melting Pt: 32…33 °C
Storage Temperature: Ambient
MDL Number: MFCD03094661
CAS Number: 53207-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR