5-Bromo-2-iodoanisole 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Bromo-1-iodo-2-methoxybenzene

OR46223-1G OR46223-25G OR46223-5G
76584-974EA 196 USD
76584-974 76584-972 76584-970
5-Bromo-2-iodoanisole 97%
5-Bromo-2-iodoanisole
Formula: C₇H₆BrIO
MW: 312.9300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847482
CAS Number: 791642-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR