2-Bromo-4-iodoanisole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-4-iodo-1-methoxybenzene

OR45120-1G OR45120-5G
76584-242EA 164 USD
76584-242 76584-240
2-Bromo-4-iodoanisole
2-Bromo-4-iodoanisole
Formula: C₇H₆BrIO
MW: 312.9304 g/mol
Melting Pt: 89…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425935
CAS Number: 182056-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR