2,4-Dichloroquinazoline 97%

Supplier: APOLLO SCIENTIFIC

Warning

OR52131-100G OR52131-25G OR52131-5G
76587-000EA 66 USD
76587-000 76587-002 76586-998
2,4-Dichloroquinazoline 97%
2,4-Dichloroquinazoline
Formula: C₈H₄Cl₂N₂
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00091950
CAS Number: 607-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR