5,6-Dibromo-2,1,3-benzothiadiazole

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 5,6-Dibromobenzo[c][1,2,5]thiadiazole

OR50963-5G OR50963-25G
76586-322EA 479 USD
76586-322 76586-320
5,6-Dibromo-2,1,3-benzothiadiazole
5,6-Dibromo-2,1,3-benzothiadiazole
Formula: C₆H₂Br₂N₂S
MW: 293.9700 g/mol
Melting Pt: 132…133 °C
Storage Temperature: Ambient
CAS Number: 18392-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR