2-Chloro-6-methylnicotinic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5068-5G OR5068-25G
76586-258EA 16.48 USD
76586-258 76586-260
2-Chloro-6-methylnicotinic acid 98%
2-Chloro-6-methylnicotinic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 157…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00134165
CAS Number: 30529-70-5
EINECS: 250-229-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR