4,7-Dibromo-3-hydroxy-2-naphthoic acid 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1,6-Dibromo-2-hydroxynaphthalene-3-carboxylic acid

OR7538-100G OR7538-5G OR7538-25G
76588-028EA 101 USD
76588-028 76588-026 76588-024
4,7-Dibromo-3-hydroxy-2-naphthoic acid 97%
4,7-Dibromo-3-hydroxy-2-naphthoic acid
Formula: C₁₁H₆Br₂O₃
Melting Pt: 251…255 °C
Storage Temperature: Ambient
MDL Number: MFCD00046367
CAS Number: 1779-10-8
EINECS: 217-214-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR