Methyl-4-ethynylbenzoate 96%

Supplier: APOLLO SCIENTIFIC

Synonyms: Methyl 4-ethynylbenzoate

OR48275-25G OR48275-1G OR48275-5G
76585-830EA 265 USD
76585-830 76585-828 76585-826
Methyl-4-ethynylbenzoate 96%
Methyl 4-ethynylbenzoate
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00168820
CAS Number: 3034-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR