Imidazole-4,5-dicarboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4,5-Imidazoledicarboxylic acid1H-Imidazole-4,5-dicarboxylic acid

OR43522-25G OR43522-5G OR43522-100G
76583-790EA 88.58 USD
76583-790 76583-786 76583-788
Imidazole-4,5-dicarboxylic acid
Imidazole-4,5-dicarboxylic acid
Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
Melting Pt: 280…285 °C
Storage Temperature: Ambient
MDL Number: MFCD00005200
CAS Number: 570-22-9
EINECS: 209-327-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR