(-)-Dibenzoyl-L-tartaric acid monohydrate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate

OR8726-5KG OR8726-250G OR8726-1KG
76589-024EA 259 USD
76589-024 76589-020 76589-022
(-)-Dibenzoyl-L-tartaric acid monohydrate
(-)-Dibenzoyl-L-tartaric acid monohydrate

Optical resolving agent

Formula: C₁₈H₁₄O₈·1H₂O
MW: 376.33 (358.31 anhy)
Melting Pt: 88…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00149119
CAS Number: 62708-56-9
EINECS: 613-072-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR