2-Methyl-1-tetralone 99%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Methyl-3,4-dihydronaphthalen-1(2H)-one

OR46762-5G OR46762-1G OR46762-25G
76585-248EA 137 USD
76585-248 76585-246 76585-250
2-Methyl-1-tetralone 99%
2-Methyl-1-tetralone
Formula: C₁₁H₁₂O
MW: 160.22 g/mol
Boiling Pt: 127…131 °C/12mmHg
Density: 1.057 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001690
CAS Number: 1590-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR