(±)-1,2,3,4-Tetrahydro-2-naphthol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,2,3,4-Tetrahydronaphthalen-2-ol

OR471677-1G
76585-420EA 38 USD
76585-420
(±)-1,2,3,4-Tetrahydro-2-naphthol 98%
(±)-1,2,3,4-Tetrahydro-2-naphthol
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045575
CAS Number: 530-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR