2-Hydroxyhippuric acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-(2-Hydroxybenzoyl)glycine

OR4276-250MG OR4276-1G
76583-676EA 23 USD
76583-676 76583-678
2-Hydroxyhippuric acid
2-Hydroxyhippuric acid
Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002695
EINECS: 207-661-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR