5-Bromo-2-methoxypyrimidine 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR7810-100G OR7810-25G
76588-288EA 202 USD
76588-288 76588-286
5-Bromo-2-methoxypyrimidine 98%
5-Bromo-2-methoxypyrimidine
Formula: C₅H₅BrN₂O
Melting Pt: 55…60 °C
Storage Temperature: Ambient
MDL Number: MFCD04973351
CAS Number: 14001-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR