5-Methylhydantoin 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR451305-1G OR451305-25G OR451305-5G
76584-280EA 404 USD
76584-280 76584-278 76584-276
5-Methylhydantoin 95%
5-Methylhydantoin
Formula: C₄H₆N₂O₂
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00022398
CAS Number: 616-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR