Ethyl picolinate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Ethyl 2-pyridinecarboxylateEthyl 2-picolinate

OR4533-10G OR4533-100G
76584-720EA 57 USD
76584-720 76584-718
Ethyl picolinate
Ethyl picolinate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 240…241 °C
Melting Pt: 44256 °C
Density: 1.119 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00006292
CAS Number: 2524-52-9
EINECS: 219-758-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR