3-Bromo-5-formylpyridine 97+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Bromonicotinaldehyde5-Bromo-3-pyridinecarboxaldehyde3-Bromopyridine-5-carboxaldehyde5-Bromopyridine-3-carboxaldehyde

OR4712-25G OR4712-1G OR4712-5G
76585-388EA 51 USD
76585-388 76585-390 76585-392
3-Bromo-5-formylpyridine 97+%
3-Bromo-5-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD03265758
CAS Number: 113118-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR