4-Chloropyridine hydrochloride 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Chloropyridinium chloride

OR6949-5G OR6949-100G OR6949-25G
76587-286EA 54 USD
76587-286 76587-288 76587-284
4-Chloropyridine hydrochloride 97%
4-Chloropyridine hydrochloride
Formula: C₅H₅Cl₂N
Melting Pt: 210 °C (subl.)
Flash Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00012829
CAS Number: 7379-35-3
EINECS: 230-946-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR