(±)-Tropic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Phenylhydracrylic acid3-Hydroxy-2-phenylpropionic acidTropic acidDL-Tropic acid

OR4935-25G OR4935-100G OR4935-500G
76585-930EA 54 USD
76585-930 76585-932 76585-928
(±)-Tropic acid 98%
(±)-Tropic acid
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00004255
CAS Number: 529-64-6
EINECS: 209-020-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR