1-Pentylpiperazine

Supplier: APOLLO SCIENTIFIC

Warning

OR4168-1G OR4168-250MG
76583-310EA 22 USD
76583-310 76583-312
1-Pentylpiperazine
1-Pentylpiperazine
Formula: C₉H₂₀N₂
MW: 156.27 g/mol
Boiling Pt: 88…91 °C/20mmHg
Storage Temperature: Ambient
MDL Number: MFCD02093555
CAS Number: 50866-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR