4-Nitrophenethyl bromide

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-(2-Bromoethyl)-4-nitrobenzene2-(4-Nitrophenyl)-1-bromoethane

OR8248-50G OR8248-10G
76588-658EA 31 USD
76588-658 76588-656
4-Nitrophenethyl bromide
4-Nitrophenethyl bromide
Formula: C₈H₈BrNO₂
Boiling Pt: 116 °C/0.2mmHg
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00007386
CAS Number: 5339-26-4
EINECS: 226-271-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR