3-Benzyloxyaniline 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR8289-1G OR8289-10G OR8289-5G
76588-690EA 43 USD
76588-690 76588-688 76588-692
3-Benzyloxyaniline 98%
3-Benzyloxyaniline
Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00007784
CAS Number: 1484-26-0
EINECS: 216-056-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR