4-Acetoxybenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR7979-250G OR7979-50G OR7979-500G
76588-476EA 133 USD
76588-476 76588-474 76588-472
4-Acetoxybenzoic acid
4-Acetoxybenzoic acid
Formula: C₉H₈O₄
MW: 180.16 g/mol
Boiling Pt: 100…120 °C/20mmHg
Melting Pt: 190…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00002540
CAS Number: 2345-34-8
EINECS: 219-066-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR