2-Chloro-3-methoxybenzonitrile ≥98%

Supplier: APOLLO SCIENTIFIC

OR510010-1G
76586-400EA 105 USD
76586-400
2-Chloro-3-methoxybenzonitrile ≥98%
2-Chloro-3-methoxybenzonitrile
Formula: C₈H₆ClNO
MW: 167.59 g/mol
Storage Temperature: Refrigerator
CAS Number: 853331-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR