2-Bromo-5-nitro-6-picoline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-6-methyl-5-nitropyridine6-Bromo-2-methyl-3-nitropyridine

OR8428-5G OR8428-25G
76588-788EA 27 USD
76588-788 76588-786
2-Bromo-5-nitro-6-picoline
2-Bromo-5-nitro-6-picoline
Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Melting Pt: 69…70 °C
Storage Temperature: Ambient
MDL Number: MFCD03095219
CAS Number: 22282-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR