5-Quinolinecarboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

OR451147-5G OR451147-1G
76584-204EA 90 USD
76584-204 76584-202
5-Quinolinecarboxylic acid
5-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047608
CAS Number: 7250-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR