8-Bromoquinoline 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR7885-25G OR7885-5G
76588-370EA 55 USD
76588-370 76588-368
8-Bromoquinoline 98%
8-Bromoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 112…113 °C/0.5mmHg
Density: 1.594 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00191859
CAS Number: 16567-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR