3,5-Dimethylbenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR4988-100G OR4988-500G OR4988-1KG
76585-982EA 46 USD
76585-982 76585-984 76585-980
3,5-Dimethylbenzoic acid 98%
3,5-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00002525
CAS Number: 499-06-9
EINECS: 207-876-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR