5-Nitro-o-toluic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR7737-100G OR7737-10G
76588-196EA 118 USD
76588-196 76588-194
5-Nitro-o-toluic acid 98%
5-Nitro-o-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 177…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00007371
CAS Number: 1975-52-6
EINECS: 217-829-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR