Methyl-3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate 95%

Supplier: APOLLO SCIENTIFIC

Warning

PC9397-1G
76562-662EA 86 USD
76562-662
Methyl-3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate 95%
Methyl-3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate
Formula: C₁₂H₉ClFNO₃
MW: 269.66 g/mol
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00214743
CAS Number: 4415-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR