Methyl-3-(2-chloro-6-fluorophenyl)-5-[2-(dimethylamino)vinyl]-4-isoxazolecarboxylate

Supplier: APOLLO SCIENTIFIC

Warning

PC9395-250MG PC9395-1G
76560-622EA 58 USD
76560-622 76560-754
Methyl-3-(2-chloro-6-fluorophenyl)-5-[2-(dimethylamino)vinyl]-4-isoxazolecarboxylate
Methyl-3-(2-chloro-6-fluorophenyl)-5-[2-(dimethylamino)vinyl]-4-isoxazolecarboxylate
Formula: C₁₅H₁₄ClFN₂O₃
MW: 324.74 g/mol
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00127467
CAS Number: 338417-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR