N-Ethyl-2-nitro-4-(trifluoromethyl)aniline

Supplier: APOLLO SCIENTIFIC

Warning

PC8794-1G PC8794-250MG PC8794-5G
76561-556EA 37 USD
76561-556 76563-352 76560-136
N-Ethyl-2-nitro-4-(trifluoromethyl)aniline
N-Ethyl-2-nitro-4-(trifluoromethyl)aniline
Formula: C₉H₉F₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00044178
CAS Number: 30377-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR